Coated slides
Lieferant: Merck
703176-10PAK
SIAL703176-10PAKEA 79.6 EUR
SIAL703176-10PAK
Coated slides
Objektträger Adhäsions-Objektträger
Indium tin oxide coated glass slides.
  • Good resistivity of 70-100 Ω/sq
  • Unpolished surface with 86 % transmittance