3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

703265-1G
SIAL703265-1GEA 73 EUR
SIAL703265-1G
3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbaldehyd, Sigma-Aldrich®
3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbaldehyd
Formel: C₁₁H₈FNO₂
Molecular Weight: 205,19 g/mol
MDL: MFCD11656172
CAS-Nummer: 869496-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR