3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

703273-1G
SIAL703273-1GEA 32.6 EUR
SIAL703273-1G
3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbaldehyd, Sigma-Aldrich®
3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbaldehyd
Formel: C₁₂H₁₁NO₃
MDL: MFCD12196085
CAS-Nummer: 273223-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR