Ethylmagnesiumbromid 3.4 mol/l, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
703591-100ML
703591-800ML
SIAL703591-100MLEA
282
EUR
SIAL703591-100ML
SIAL703591-800ML
Ethylmagnesiumbromid 3.4 mol/l, Sigma-Aldrich®
Ethylmagnesium bromide
Formel:
C₂H₅BrMg Molecular Weight: 133.27 g/mol |
MDL:
MFCD00000043 CAS-Nummer: 925-90-6 EINECS: 213-127-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...