Ethylmagnesiumbromid 3.4 mol/l, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

703591-100ML 703591-800ML
SIAL703591-100MLEA 282 EUR
SIAL703591-100ML SIAL703591-800ML
Ethylmagnesiumbromid 3.4 mol/l, Sigma-Aldrich®
Ethylmagnesium bromide
Formel: C₂H₅BrMg
MDL: MFCD00000043
CAS-Nummer: 925-90-6
EINECS: 213-127-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR