3-Acetyl-5-brompyridin, Sigma-Aldrich®

Lieferant: Merck

703893-1G
SIAL703893-1GEA 150 EUR
SIAL703893-1G
3-Acetyl-5-brompyridin, Sigma-Aldrich®
3-Acetyl-5-brompyridin
Formel: C₇H₆BrNO
Molecular Weight: 200,03 g/mol
MDL: MFCD03086033
CAS-Nummer: 38940-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR