3,5-Difluorpyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
703923-5G
SIAL703923-5GEA
138
EUR
SIAL703923-5G
3,5-Difluorpyridin, Sigma-Aldrich®
3,5-Difluoropyridine
Formel:
C₅H₃F₂N Molecular Weight: 115,08 g/mol Siedepunkt: 92…93 °C (1013 hPa) Dichte: 1,268 g/cm³ (20 °C) |
MDL:
MFCD03412222 CAS-Nummer: 71902-33-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...