3-Brom-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-2-pyridincarbonsäurenitril, 3-Brom-2-cyanpyridin

M-Clarity™ Qualitätsstufe = MQ100

703966-1G
SIAL703966-1GEA 70.4 EUR
SIAL703966-1G
3-Brom-2-pyridincarbonitril, Sigma-Aldrich®
3-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
MDL: MFCD02683288
CAS-Nummer: 55758-02-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR