5-Fluor-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Cyan-5-fluorpyridin, 5-Fluor-2-pyridincarbonsäurenitril

M-Clarity™ Qualitätsstufe = MQ100

703982-5G
SIAL703982-5GEA 96.7 EUR
SIAL703982-5G
5-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
5-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
Molecular Weight: 122.1 g/mol
MDL: MFCD08741371
CAS-Nummer: 327056-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR