2-Bromisonicotinsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
703990-5G
SIAL703990-5GEA
106
EUR
SIAL703990-5G
2-Bromisonicotinsäure, Sigma-Aldrich®
2-Bromisonicotinsäure
Formel:
C₆H₄BrNO₂ Molecular Weight: 202.01 g/mol |
MDL:
MFCD01646069 CAS-Nummer: 66572-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...