2-Fluorisonicotinonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluor-4-pyridincarbonsäurenitril, 4-Cyan-2-fluorpyridin

M-Clarity™ Qualitätsstufe = MQ100

704016-5G
SIAL704016-5GEA 45.3 EUR
SIAL704016-5G
2-Fluorisonicotinonitril, Sigma-Aldrich®
2-Fluorisonicotinonitril
Formel: C₆H₃FN₂
Molecular Weight: 122.1 g/mol
MDL: MFCD09027299
CAS-Nummer: 3939-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR