6-Fluor-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluor-2-pyridincarbonsäurenitril, 2-Cyan-6-fluorpyridin

M-Clarity™ Qualitätsstufe = MQ100

704024-1G
SIAL704024-1GEA 89.1 EUR
SIAL704024-1G
6-Fluor-2-pyridincarbonitril, Sigma-Aldrich®
6-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
Molecular Weight: 122.1 g/mol
MDL: MFCD09263323
CAS-Nummer: 3939-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR