2-Amino-5-brom-3-iodpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-3-iodpyridin-2-amin, 5-Brom-3-iod-2-pyridylamin

M-Clarity™ Qualitätsstufe = MQ100

704032-1G
SIAL704032-1GEA 85.8 EUR
SIAL704032-1G
2-Amino-5-brom-3-iodpyridin, Sigma-Aldrich®
2-Amino-5-brom-3-iodpyridin
Formel: C₅H₄BrIN₂
Molecular Weight: 298,91 g/mol
MDL: MFCD06659000
CAS-Nummer: 381233-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR