2-Amino-3-iodpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

704474-5G
SIAL704474-5GEA 67.6 EUR
SIAL704474-5G
2-Amino-3-iodpyridin, Sigma-Aldrich®
2-Amino-3-iodpyridin
Formel: C₅H₅IN₂
Molecular Weight: 220,01 g/mol
MDL: MFCD01651885
CAS-Nummer: 104830-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR