3-Fluor-4-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Fluoropyridine-4-boronic acid pinacol ester

704555-1G
SIAL704555-1GEA 100 EUR
SIAL704555-1G
3-Fluor-4-pyridinboronsäurepinakolester, Sigma-Aldrich®
3-Fluor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BFNO₂
Molecular Weight: 223,05 g/mol
MDL: MFCD06798251
CAS-Nummer: 458532-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR