2-Carboxyphenylboronsäure Dihydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
704784-1G
SIAL704784-1GEA
131
EUR
SIAL704784-1G
2-Carboxyphenylboronsäure Dihydrat, Sigma-Aldrich®
2-Carboxyphenylboronsäure Dihydrat
Formel:
C₇H₇BO₄·2H₂O |
MDL:
MFCD15141948 CAS-Nummer: 1257650-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...