Naphthol AS-E-phosphat (KG-501), Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(4-Chlorphenyl)-3-(phosphonooxy)naphthalin-2-carboxamid

M-Clarity™ Qualitätsstufe = MQ100

70485-250MG
SIAL70485-250MGEA 72.6 EUR
SIAL70485-250MG
Naphthol AS-E-phosphat (KG-501), Sigma-Aldrich®
Naphthol AS-E-phosphat (KG-501)
Formel: C₁₇H₁₃ClNO₅P
Molecular Weight: 377,72 g/mol
CAS-Nummer: 18228-17-6
EINECS: 242-106-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR