2-Brom-3-iodpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

704857-1G
SIAL704857-1GEA 90.6 EUR
SIAL704857-1G
2-Brom-3-iodpyridin, Sigma-Aldrich®
2-Bromo-3-iodopyridine
Formel: C₅H₃BrIN
Molecular Weight: 283,89 g/mol
MDL: MFCD09056782
CAS-Nummer: 265981-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR