Tetraethylenglycolmonomethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: mPEG4-OH

M-Clarity™ Qualitätsstufe = MQ100

705276-25G
SIAL705276-25GEA 246 EUR
SIAL705276-25G
Tetraethylenglycolmonomethylether, Sigma-Aldrich®
Tetraethylenglycolmonomethylether
Formel: C₉H₂₀O₅
Molecular Weight: 208,25 g/mol
MDL: MFCD00041756
CAS-Nummer: 23783-42-8
EINECS: 245-883-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR