Chlorwasserstoff 3 mol/l in Cyclopentylmethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chlorwasserstoff

705551-100ML
SIAL705551-100MLEA 87.3 EUR
SIAL705551-100ML
Chlorwasserstoff 3 mol/l in Cyclopentylmethylether, Sigma-Aldrich®
Chlorwasserstoff
Formel: HCl
Molecular Weight: 36.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR