2,6-Dimethoxy-3-pyridinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

706086-250MG
SIAL706086-250MGEA 61.2 EUR
SIAL706086-250MG
2,6-Dimethoxy-3-pyridinboronsäure, Sigma-Aldrich®
2,6-Dimethoxy-3-pyridineboronic acid
Formel: C₇H₁₀BNO₄
Molecular Weight: 182.97 g/mol
MDL: MFCD03788239
CAS-Nummer: 221006-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR