4-(Furfurylcarbamoyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

706183-5G
SIAL706183-5GEA 270 EUR
SIAL706183-5G
4-(Furfurylcarbamoyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(Furfurylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₂BNO₄
MDL: MFCD09266168
CAS-Nummer: 1073353-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR