2,5-Dichlorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dichloropyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

706299-1G
SIAL706299-1GEA 49.2 EUR
SIAL706299-1G
2,5-Dichlorisonicotinsäure, Sigma-Aldrich®
2,5-Dichlorisonicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD00466640
CAS-Nummer: 88912-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR