4-Brom-3-fluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

706310-1G
SIAL706310-1GEA 70.6 EUR
SIAL706310-1G
4-Brom-3-fluorbenzaldehyd, Sigma-Aldrich®
4-Bromo-3-fluorobenzaldehyde
Formel: C₇H₄BrFO
Molecular Weight: 203,01 g/mol
MDL: MFCD03095000
CAS-Nummer: 133059-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR