4,6-Dichlor-5-methoxypyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

706485-1G
SIAL706485-1GEA 69.9 EUR
SIAL706485-1G
4,6-Dichlor-5-methoxypyrimidin, Sigma-Aldrich®
4,6-Dichloro-5-methoxypyrimidine
Formel: C₅H₄Cl₂N₂O
Molecular Weight: 179,01 g/mol
MDL: MFCD00087680
CAS-Nummer: 5018-38-2
EINECS: 225-699-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR