(±)-1-Boc-3-aminopyrrolidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine

M-Clarity™ Qualitätsstufe = MQ100

706582-1G
SIAL706582-1GEA 55.7 EUR
SIAL706582-1G
(±)-1-Boc-3-aminopyrrolidin, Sigma-Aldrich®
(±)-1-Boc-3-aminopyrrolidin
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186,25 g/mol
MDL: MFCD01861220
CAS-Nummer: 186550-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR