3-Acetyl-2-brompyridin, Sigma-Aldrich®

Lieferant: Merck

706701-250MG
SIAL706701-250MGEA 37 EUR
SIAL706701-250MG
3-Acetyl-2-brompyridin, Sigma-Aldrich®
3-Acetyl-2-brompyridin
Formel: C₇H₆BrNO
Molecular Weight: 200,0300 g/mol
MDL: MFCD04114370
CAS-Nummer: 84199-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR