4-Brom-2,6-difluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

706957-5G
SIAL706957-5GEA 97.6 EUR
SIAL706957-5G
4-Brom-2,6-difluorbenzaldehyd, Sigma-Aldrich®
4-Bromo-2,6-difluorobenzaldehyde
Formel: C₇H₃BrF₂O
Molecular Weight: 221 g/mol
MDL: MFCD03094459
CAS-Nummer: 537013-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR