Ethylthiazol-4-carboxylat, Sigma-Aldrich®

Lieferant: Merck

707112-500MG
SIAL707112-500MGEA 28.9 EUR
SIAL707112-500MG
Ethylthiazol-4-carboxylat, Sigma-Aldrich®
Ethylthiazol-4-carboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.1900 g/mol
MDL: MFCD03788566
CAS-Nummer: 14527-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR