2-Amino-3-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

707341-1G
SIAL707341-1GEA 66.5 EUR
SIAL707341-1G
2-Amino-3-fluorpyridin, Sigma-Aldrich®
2-Amino-3-fluorpyridin
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
MDL: MFCD04114135
CAS-Nummer: 21717-95-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR