1H-Pyrazol-3-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

707384-1G
SIAL707384-1GEA 32.7 EUR
SIAL707384-1G
1H-Pyrazol-3-carbonsäure, Sigma-Aldrich®
1H-Pyrazole-3-carboxylic acid
Formel: C₄H₄N₂O₂
Molecular Weight: 112,09 g/mol
MDL: MFCD00077436
CAS-Nummer: 1621-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR