2,5-Dibrom-4-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dibrom-4-methylpyridin

708143-1G
SIAL708143-1GEA 56 EUR
SIAL708143-1G
2,5-Dibrom-4-picolin, Sigma-Aldrich®
2,5-Dibrom-4-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
MDL: MFCD00234955
CAS-Nummer: 3430-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR