1,1-Cyclohexandicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyclohexane-1,1-dicarboxylic acid

708216-1G
SIAL708216-1GEA 46.8 EUR
SIAL708216-1G
1,1-Cyclohexandicarbonsäure, Sigma-Aldrich®
1,1-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
Molecular Weight: 172,18 g/mol
MDL: MFCD00094668
CAS-Nummer: 1127-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR