1-Butyl-2,3-dimethylimidazoliumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate, [BDiMIM] [BF4]

70863-50G-F
SIAL70863-50G-FEA 242 EUR
SIAL70863-50G-F
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborat, Sigma-Aldrich®
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Formel: C₉H₁₇BF₄N₂
Molecular Weight: 240,05 g/mol
Dichte: 1,2 g/cm³ (20 °C)
MDL: MFCD03427618
CAS-Nummer: 402846-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR