6-Chinoxalincarbonsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Quinoxaline-6-boronic acid, pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

708631-1G
SIAL708631-1GEA 165 EUR
SIAL708631-1G
6-Chinoxalincarbonsäurepinakolester, Sigma-Aldrich®
6-Chinoxalincarbonsäurepinakolester
Formel: C₁₄H₁₇BN₂O₂
Molecular Weight: 256.1080 g/mol
MDL: MFCD11054040
CAS-Nummer: 1167418-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR