1-Butyl-2,3-dimethylimidazoliumhexafluorophosphat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Butyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)

70869-5G-F
SIAL70869-5G-FEA 68 EUR
SIAL70869-5G-F
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphat, Sigma-Aldrich®
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate
Formel: C₉H₁₇F₆N₂P
Molecular Weight: 298,21 g/mol
MDL: MFCD03790877
CAS-Nummer: 227617-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR