Eisen(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

708801-1G
SIAL708801-1GEA 71.2 EUR
SIAL708801-1G
Eisen(III)trifluormethansulfonat, Sigma-Aldrich®
Eisen(III)trifluormethansulfonat
Formel: C₃F₉FeO₉S₃
Molecular Weight: 503,06 g/mol
MDL: MFCD15144785
CAS-Nummer: 63295-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR