2-Methoxyethylisothiocyanat, Sigma-Aldrich®
Lieferant: Merck
709123-5G
SIAL709123-5GEA
126
EUR
SIAL709123-5G
2-Methoxyethylisothiocyanat, Sigma-Aldrich®
2-Methoxyethylisothiocyanat
Formel:
C₄H₇NOS Molecular Weight: 117.17 g/mol Siedepunkt: 88.5…92 °C (23.5 mmHg) Dichte: 1.08 g/cm³ (20 °C) |
MDL:
MFCD00041126 CAS-Nummer: 38663-85-3 EINECS: 254-070-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...