2-Amino-4-(trifluormethyl)pyrimidin, Sigma-Aldrich®

Lieferant: Merck

709131-1G
SIAL709131-1GEA 55.4 EUR
SIAL709131-1G
2-Amino-4-(trifluormethyl)pyrimidin, Sigma-Aldrich®
2-Amino-4-(trifluoromethyl)pyrimidine
Formel: C₅H₄F₃N₃
Molecular Weight: 163,1 g/mol
MDL: MFCD00828624
CAS-Nummer: 16075-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR