(S)-(-)-1-(2-Naphthyl)ethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-1-(Naphthalen-2-yl)ethanamine

70942-1G
SIAL70942-1GEA 140 EUR
SIAL70942-1G
(S)-(-)-1-(2-Naphthyl)ethylamin, Sigma-Aldrich®
(S)-(-)-1-(2-Naphthyl)ethylamin
Formel: C₁₂H₁₃N
Molecular Weight: 171,24 g/mol
MDL: MFCD00085366
CAS-Nummer: 3082-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR