3-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-((Trimethylsilyl)ethynyl)benzaldehyde
709727-250MG
SIAL709727-250MGEA
63.7
EUR
SIAL709727-250MG
3-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
3-[(Trimethylsilyl)ethinyl]benzaldehyd
Formel:
C₁₂H₁₄OSi Molecular Weight: 202.324 g/mol |
MDL:
MFCD06245348 CAS-Nummer: 77123-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...