2-Brom-5-hydroxypyrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromopyrazin-2-ol

709735-250MG
SIAL709735-250MGEA 51.6 EUR
SIAL709735-250MG
2-Brom-5-hydroxypyrazin, Sigma-Aldrich®
2-Bromo-5-hydroxypyrazine
Formel: C₄H₃BrN₂O
Molecular Weight: 174.9800 g/mol
MDL: MFCD06245330
CAS-Nummer: 374063-92-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR