4-Methylthiobenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thio-p-toluamid, 4-Methylthiobenzoesäureamid

709786-1G
SIAL709786-1GEA 41.6 EUR
SIAL709786-1G
4-Methylthiobenzamid, Sigma-Aldrich®
4-Methylthiobenzamid
Formel: C₈H₉NS
Molecular Weight: 151,23 g/mol
MDL: MFCD00173750
CAS-Nummer: 2362-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR