2-Methylbenzamidoxim, Sigma-Aldrich®

Lieferant: Merck

709794-1G
SIAL709794-1GEA 56 EUR
SIAL709794-1G
2-Methylbenzamidoxim, Sigma-Aldrich®
2-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
CAS-Nummer: 40312-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR