2-Brom-6-fluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

709824-1G
SIAL709824-1GEA 22.5 EUR
SIAL709824-1G
2-Brom-6-fluorbenzaldehyd, Sigma-Aldrich®
2-Bromo-6-fluorobenzaldehyde
Formel: C₇H₄BrFO
Molecular Weight: 203,01 g/mol
MDL: MFCD03407341
CAS-Nummer: 360575-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR