3-Fluor-2-formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid

709832-1G
SIAL709832-1GEA 57.3 EUR
SIAL709832-1G
3-Fluor-2-formylphenylboronsäure, Sigma-Aldrich®
3-Fluor-2-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,93 g/mol
MDL: MFCD10697421
CAS-Nummer: 871126-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR