Chlorbis(3,5-dimethyl-4-methoxyphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

710385-1G
SIAL710385-1GEA 151 EUR
SIAL710385-1G
Chlorbis(3,5-dimethyl-4-methoxyphenyl)phosphin, Sigma-Aldrich®
Chlorbis(3,5-dimethyl-4-methoxyphenyl)phosphin
Formel: C₁₈H₂₂ClO₂P
MDL: MFCD01630810
CAS-Nummer: 136802-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR