5-Bromisophthalsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-1,3-benzoldicarbonsäure

711217-5G
SIAL711217-5GEA 35.4 EUR
SIAL711217-5G
5-Bromisophthalsäure, Sigma-Aldrich®
5-Bromisophthalsäure
Formel: C₈H₅BrO₄
Molecular Weight: 245,03 g/mol
MDL: MFCD00152019
CAS-Nummer: 23351-91-9
EINECS: 245-602-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR