2',4',5'-Trimethoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

711233-25G
SIAL711233-25GEA 99.1 EUR
SIAL711233-25G
2',4',5'-Trimethoxyacetophenon, Sigma-Aldrich®
2',4',5'-Trimethoxyacetophenon
Formel: C₁₁H₁₄O₄
MDL: MFCD00017237
CAS-Nummer: 1818-28-6
EINECS: 217-333-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR