4-Aminotetrahydropyran, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrahydro-2H-pyran-4-amine

711357-1G
SIAL711357-1GEA 143 EUR
SIAL711357-1G
4-Aminotetrahydropyran, Sigma-Aldrich®
4-Aminotetrahydropyran
Formel: C₅H₁₁NO
Molecular Weight: 101.1469 g/mol
MDL: MFCD02179436
CAS-Nummer: 38041-19-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR